DuPont ™ Suprel ™ LS Lab Coats
Comfortable and breathable
Manufacturer: Dupont Personal Protection SR219SWH3X003000
 View all options for this product
Catalog No. 19-165-186
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Color White
    Size 3X-Large
    Offer a superior balance of breathability and barrier protection combined with a softer feel and increased comfort.
    • Mandarin collar
    • Knit cuffs
    • Three pockets
    • Six-snap front closure
    • Serged seams
    • Color: White
    Suprel LS is not flame-resistant. Should not be used around heat, flame, sparks, or in potential flammable or explosive environments.
  • Description & Specifications
Related Products
Product Content Feedback