Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 30 Boot Covers

Catalog Number 19080722C
Click to view available options
Size:
Large
X-Large
Length (English):
15 in.
17 in.
Length (Metric):
38.1 cm
43.18 cm
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.

Boot covers are 13 in. high with elastic top for secure fit and are suitable for controlled envnironments.

  • Boot covers protects wearers and the controlled environment
  • Protects against nonhazardous liquids and dry particulates
  • Spunbonded polypropylene with polyethylene film coating
  • High slip resistance, wet and dry
  • Resists delamination, linting
  • Serged seams
  • Elastic ankles for secure fit
  • 33cm (13 in.) height with elastic top
TRUSTED_SUSTAINABILITY
Size Large
Material Spunbond Polypropylene
Latex Free Yes
Coating Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Seam Type Serged
Certifications/Compliance ASTM D3776
Packaging Quantity 100 Pairs/Cs.
Packaging Type Bulk Packaged, Double Transparent Poly Liners
Length (English) 15 in.
Length (Metric) 38.1 cm
Height (English) 13 in.
Height (Metric) 33.02 cm
Resistance Dry Particulates, Splash
Product Type Controlled Environments Boot Covers
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit