Learn More
DuPont™ ProShield 30™ Shoe Covers
Help to keep workers clean and safe
Manufacturer:  DuPont™ PE440SWHXL0200CT
 View more versions of this product
Catalog No. 19-080-721D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Help to keep workers clean and safe
  • Resistant to liquids
  • Will not delaminate
  • Lint-free
  • Generously sized
  • Have skid-resistant sole
  • Elastic ankles for secure fit
  • Seamless
Specifications
Specifications
Safety
Polyethylene extrusion coating
X-Large
Ideal to keep workers clean and safe
13.97cm
Yes
Serged
Elastic
White
Spunbond Polypropylene
5.5 in.
16.5 in.
41.91cm