Learn More
DuPont™ Tempro™ Flame-Retardant Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
Size (Metric) Inseam:
71.12 cm
73.66 cm
74.93 cm
77.47 cm
80 cm
82.55 cm
87.6 cm
This item is not returnable. View return policy
Not suitable for liquid and dry chemical protection.
This item is not returnable. View return policy
Not suitable for liquid and dry chemical protection.
Provide enhanced protection from moisture, sparks and flames for applications in steel mills, coal mines, refineries, laboratories, and by utilities workers. Made of nonwoven fabric wood-pulp/polyester blend treated to provide fire retardant and liquid repellancy. Tempro coveralls are antistatic and breathable and can be worn over primary thermal protective garments as a first line of defense for wearer and thermal garments underneath. Disposable protection against nuisance contamination by solids and nontoxic liquids. Front zipper and collar with serged seams for added protection. No elastic. Set sleeve for greater arm movement. Light blue. Case of 25.
Size 6X-Large
Color Blue
Closure Type Zipper
Collar Type Lay down
Size (English) Inseam 34.5 in.
Size (Metric) Inseam 87.6 cm