Learn More
DuPont™ Tempro™ TM 127S Coveralls
Offer an extra layer of protection for primary fire-resistant clothing
Manufacturer:  DuPont™ TM127SBU4X002500
 View more versions of this product
Catalog No. 19-054-006
Add to cart
Alert:
Should not be worn in presence of hazardous materials.
Description
Description
Worn over flame-resistant clothing to provide a barrier to nonhazardous particles and aerosols. Fit easily over fire-resistant clothing.
  • Elastic wrists and ankles
  • Attached hood with elastic around face
  • Front zipper closure
  • Serged seam
  • Nonwoven wood-pulp/polyester, flame-retardant, liquid repellant fabric
  • Protect and preserve primary flame-resistant clothing
  • Disposable
Can be used in steel mills, coal mines, maintenance, refineries, utilities, laboratories
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
Specifications
Elastic
Zipper
Elastic
Yes
6.4 to 6.7 ft.
Standard fit
100.33cm
Hooded
4X-Large
128.9 to 137.79cm
Safety
Blue
Non-woven Wood Pulp, Polyester Fabric
Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
1.95 to 2.04m
39.5 in.
Tempro™
Serged
32.5 in.
32 in.