DuPont ™ Tempro ™ TM 127S Coveralls

Offer an extra layer of protection for primary fire-resistant clothing

Manufacturer: Dupont Personal Protection TM127SBUMD002500

 View more versions of this product

Catalog No. 19-175-270

  Check Availability
Add to Cart

Alert

Should not be worn in presence of hazardous materials.


    Specifications

    Material Tempro™
    Seam Type Serged
    Color Blue
    Size Medium
    Disposable Yes
    Height (Metric) 1.6 to 1.73m
    For Use With (Application) Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
    Length (English) Sleeve 33.75 in.
    Length (Metric) Sleeve 85.72cm
    Closure Type Zipper
    Size (English) Inseam 28 in.
    Size (Metric) Inseam 71.12cm
    Width (English) Chest 24.25 in.
    Construction Non-woven Wood Pulp, Polyester Fabric
    Includes Standard fit hood, elastic wrist and ankle
    Size (English) Chest 35.25 to 38.75 in.
    Size (Metric) Chest 89.53 to 98.42cm
    Height (English) 5.3 to 5.7 ft.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit