Learn More
DuPont™ DuPont™ Tychem™ BR Coverall
Coverall with respirator fit hood, elastic wrists and ankles
Manufacturer:  DuPont™ BR127TYL4X000200
 View more versions of this product
Catalog No. 19-067-000
Add to cart
Description
Description
Tychem™ BR offers wide-ranging protection backed by extensive fabric permeation barrier testing. Fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents. It offers tear, puncture and abrasion resistant fabrics for lasting protection.
Specifications
Specifications
Safety
6 ft.4 in. to 6 ft.7 in.
Taped
32.62 in.
Yellow
Tychem™ BR
4X-Large