DuPont ™ Tychem ™ BR Coveralls

For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities

Manufacturer: Dupont Personal Protection BR128TYLXL0002BN

 View more versions of this product

Catalog No. 19-715-246

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance Berry Amendment
    Material Tychem™ BR
    Seam Type Taped
    Color Yellow
    Size X-Large
    Size (English) Chest 41.25 to 44.75 in.
    Height (English) 5.8 to 6.2 ft.
    Height (Metric) 1.76 to 1.88m
    For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
    Length (English) Sleeve 37.25 in.
    Length (Metric) Sleeve 94.6cm
    Closure Type Zipper Front with Storm Flap
    Size (English) Inseam 30 in.
    Size (Metric) Inseam 76.2cm
    Thickness 16mil
    Width (English) Chest 27.75 in.
    Construction Multilayer film barrier laminated to Polypropylene substrate
    Includes Respirator fit hood, elastic wrist, attached socks with outer boot flaps
    Size (Metric) Chest 107.31 to 116.2cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit