DuPont ™ Tychem ™ BR Coveralls
For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities
Manufacturer: Dupont Personal Protection BR127TYL5X000200
 View more versions of this product
Catalog No. 18-999-1139G
  Check Availability
Add to Cart
    Specifications
    Material Tychem™ BR
    Seam Type Taped
    Color Yellow
    Size 5X-Large
    For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
    Closure Type Zipper Front with Storm Flap
    Thickness 16mil
    Construction Multilayer film barrier laminated to Polypropylene substrate
  • Description & Specifications
Related Products
Product Content Feedback