Learn More
DuPont Tychem 9000 Coveralls
For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities
$100.00 - $168.00
Specifications
Color | Yellow |
---|---|
Description | Respirator fit hood, elastic wrist, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate |
Material | Tychem™ BR |
Seam Type | Taped |
For Use With (Application) | Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities |
Catalog Number | Mfr. No. | Size | Ankle Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Ankle Style | Price | Quantity & Availability | ||||
19-013-420E
|
DuPont
BR128TYL2X000200 |
2X-Large | Attached socks with outer boot flaps |
Case of 2 for $147.00
|
|
19-013-420G
|
DuPont
BR128TYL4X000200 |
4X-Large | Attached socks with outer boot flaps |
Case of 2 for $168.00
|
|
19-813-452
|
DuPont
BR128TYLLG000200 |
Large | Attached socks with outer boot flaps |
Case of 2 for $139.00
|
|
18-999-1139B
|
DuPont
BR127TYLLG000200 |
Large | Elastic |
Case of 2 for $100.00
|
|
18-999-1139A
|
DuPont
BR127TYLMD000200 |
Medium | Elastic |
Case of 2 for $100.00
|
|
18-999-1139C
|
DuPont
BR127TYLXL000200 |
X-Large | Elastic |
Case of 2 for $100.00
|
|
18-999-1139E
|
DuPont
BR127TYL3X000200 |
3X-Large | Elastic |
Case of 2 for $113.00
|
|
18-999-1139D
|
DuPont
BR127TYL2X000200 |
2X-Large | Elastic |
Case of 2 for $107.00
|
|
19-715-250
|
DuPont
BR128TYL3X000200 |
3X-Large | Attached socks with outer boot flaps |
Case of 2 for $156.00
|
|
19-715-249
|
DuPont
BR128TYLMD000200 |
Medium | Attached socks with outer boot flaps |
Case of 2 for $139.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-013-420D
|
DuPont
BR128TYLXL000200 |
X-Large | Attached socks with outer boot flaps |
Case of 2 for $139.00
|
|
Description
Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents.
- Taped seams provide strong chemical resistance against heavy liquid splashes
- A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Strong zipper for tough, flexible closure
- Elastic opening for tighter fit at wrist
- Yellow
Model BR127TYL
- Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
- Elastic opening for tighter fit at ankle
- Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Longer zipper extends to chin for complete coverage of neck area
- Integrated socks composed of garment material
Compliance
Meets ASTM D1777N, ASTM D3776, ASTM D3786, ASTM D5034; 16 CFR 1610 Standard for the Flammability of Clothing Textiles.
Specifications
Yellow | |
Tychem™ BR | |
Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities |
Respirator fit hood, elastic wrist, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate | |
Taped | |
16 mil |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.