DuPont ™ Tychem ™ BR Coveralls

For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities

Manufacturer: Dupont Personal Protection BR127TYLLG000200

 View more versions of this product

Catalog No. 18-999-1139B

  Check Availability
Add to Cart

    Specifications

    Material Tychem™ BR
    Seam Type Taped
    Color Yellow
    Size Large
    Height (Metric) 1.67 to 1.8m
    For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
    Length (Metric) Sleeve 89.5cm
    Size (English) Inseam 29.5 in.
    Thickness 16mil
    Width (English) Chest 26.37 in.
    Construction Multilayer film barrier laminated to Polypropylene substrate
    Includes Respirator fit hood, elastic wrist and ankle
    Size (Metric) Chest 99.69 to 109.22cm
    Height (English) 5.5 to 5.9 ft.
    Length (English) Sleeve 35.25 in.
    Closure Type Zipper Front with Storm Flap

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit