Learn More
DuPont ™ Tychem ™ BR Coveralls
For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities
Manufacturer:  Dupont Personal Protection BR127TYL2X000200
 View more versions of this product
Catalog No. 18-999-1139D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper Front with Storm Flap
Tychem™ BR
Elastic
Elastic
Yellow
6 to 6.4 ft.
Respirator-fit
37.87 in.
30.87 in.
29 in.
Respirator fit hood, elastic wrist and ankle; Multilayer film barrier laminated to Polypropylene substrate
113.66 to 122.55cm
Hooded
Taped
2X-Large
1.82 to 1.95m
Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
96cm
16mil
Accessories