DuPont ™ Tychem ™ BR Encapsulated Suit

Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment

Manufacturer: Dupont Personal Protection BR528TYLMD0001NY

 View more versions of this product

Catalog No. 19-101-200

  Check Availability
Add to Cart

    Specifications

    Color Yellow
    Height (English) 5 to 5.9 ft.
    Height (Metric) 1.52 to 1.79m
    For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
    Thickness 16mil
    Width (English) Chest 29.25 in.
    Material Tychem™ BR
    Includes Encapsulated level B suit, expanded back, elastic wrist, attached socks with outer boot flaps, double storm flap
    Seam Type Taped
    Size Medium
    Size (English) Chest 45.25 to 48.75 in.
    Size (Metric) Chest 114.93 to 123.82cm
    Length (English) Sleeve 28.25 in.
    Length (Metric) Sleeve 71.7cm
    Closure Type Hook and Loop
    Size (English) Inseam 29.75 in.
    Construction Multilayer film barrier laminated to Polypropylene substrate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit