Learn More
DuPont Tychem BR Splash Suit
Lightweight fabric has composite film on one side of a nonwoven substrate
$345.00 - $366.00
Specifications
Closure Type | Hook and loop with double storm flap |
---|---|
Color | Yellow |
Description | BR encapsulated level B suit, expanded back, rear entry, standard visor, 1 layer: 20 mil PVC, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure, two exhaust vents |
Material | Tychem™ BR |
Seam Type | Taped |
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
18-999-1141C
|
DuPont
BR528TYL2X000100 |
2X-Large | 6.3 to 6.5 ft. | 1.92 to 1.98 m |
Case for $366.00
|
|
18-999-1141B
|
DuPont
BR528TYLXL000100 |
X-Large | 5.8 to 6.2 ft. | 1.76 to 1.88 m |
Each for $345.00
|
|
19-130-0019
|
DuPont
BR528TYLMD000100 |
Medium | 5 to 5.9 ft. | 1.52 to 1.79 m |
Case for $345.00
|
|
Description
Lightweight, safety-yellow Tychem BR fabric is 16mil thick and weighs 6.6 oz. per sq. yd. Offers broad protection for hazardous materials handlers. Tear-, puncture-, and abrasion-resistant.
- Fully encapsulated Level B splash suit
- Large 20mil PVC faceshield
- Attached boots with outer boot flaps
- Expanded back accommodates SCBA
- Elastic wrists
- Two exhaust vents
- Zipper closure with double storm flap
- Taped seams
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications
Specifications
Hook and loop with double storm flap | |
BR encapsulated level B suit, expanded back, rear entry, standard visor, 1 layer: 20 mil PVC, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure, two exhaust vents | |
Taped | |
18 mil |
Yellow | |
Tychem™ BR | |
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.