Learn More
DuPont™ Tychem™ BR Splash Suit
Lightweight fabric has composite film on one side of a nonwoven substrate
Manufacturer:  Dupont Personal Protection BR528TYL2X000100
 View more versions of this product
Catalog No. 18-999-1141C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Double storm flap with hook and loop
BR encapsulated level B suit, expanded back, rear entry, standard visor, 1 layer: 20 mil PVC, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure, two exhaust vents
Yellow
49.75 to 53.25 in.
6.3 to 6.5 ft.
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications
78cm
82.55cm
Tychem™ BR
Taped
2X-Large
126.35 to 135.25cm
1.92 to 1.98m
30.75 in.
32.5 in.
18mil