Learn More
DuPont™ Tychem™ CPF™ 3 128 Series Coverall
DuPont Tychem 5000 Coverall. Respirator Fit Hood. Elastic Wrists. Attached Socks with Outer Boot Flaps. Storm Flap with Adhesive Closure. Taped Seams. Tan. Size 5X.
Manufacturer:  DuPont™ C3128TTN5X00062K
Catalog No. 19-715-269
Add to cart
Description
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.Attached hood with elastic around face openingStrong zipper for tough, flexible closureStorm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipperElastic opening for tighter fit at wristIntegrated socks composed of garment materialAttached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Specifications
Safety
Coverall