Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ CPF+3 360T TN Bib Overall
Click to view available options
Size:
2X-Large
3X-Large
Size (Metric) Chest:
109.85 to 118.74 cm
120 to 128.9 cm
Length (Metric):
182.88 to 195.07 cm
188.97 to 195.07 cm
Length (English) Inseam:
30.75 in.
31.5 in.
Description
- Made of a multi-layer barrier laminated to polypropylene substrate provides strength and durability
- Protection against 149 chemical situations
- Taped seams
- Bib style with adjustable webbing straps, buckle closure
- Open ankles
Specifications
Closure Type | Snap |
Color | Tan |
Material | Polypropylene |
Seam Type | Taped |
Size | 3X-Large |
Ankle Style | Elastic |
Description | Open ankle, adjustable webbing straps with buckle closure; Tychem™ CPF3 Fabric is composed of a multilayer film barrier laminated to a durable 2 oz. Polypropylene substrate |
Cuff Style | Elastic |
For Use With (Application) | C3670T TN jacket or in appplications where potential exposure is below waist. Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness |
Length (English) | 6.2 to 6.4 ft. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction