Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ DuPont™ Tychem™ CPF 3 Coverall With Collar, Elastic Wrist and Ankles

Taped seams provide strong chemical resistance against heavy liquid splashes

$328.38 - $430.47

Specifications

Material Tychem™ CPF3 fabric
Color Tan
Seam Type Taped
View More Specs

Products 6
Catalog Number Mfr. No. Size Width (English) Chest Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Width (English) Chest Height (English) Price Quantity & Availability  
19-067-009
View Documents
DuPont™
C3125TTNLG000600
Large 25.75 in. 5 ft.5 in. to 5 ft. 9 in.
Case of 6 for $328.38
Add to cart
 
19-067-010
View Documents
DuPont™
C3125TTNXL000600
X-Large 27.75 in. 5 ft. 8 in. to 6 ft. 2 in.
Case of 6 for $328.38
Add to cart
 
19-067-005
View Documents
DuPont™
C3125TTN2X000600
2X-Large 29.75 in. 6 ft. to 6 ft. 4 in.
Case of 6 for $348.37
Add to cart
 
19-067-006
View Documents
DuPont™
C3125TTN3X000600
3X-Large 31.75 in. 6 ft.2 in. to 6 ft. 4 in.
Case of 6 for $368.38
Add to cart
 
19-067-007
View Documents
DuPont™
C3125TTN4X000600
4X-Large 34 in. 6 ft. 4 in. to 6 ft. 7 in.
Case of 6 for $397.85
Add to cart
 
19-067-008
View Documents
DuPont™
C3125TTN5X000600
5X-Large 36.5 in. 6 ft. 7 in. to 6 ft. 10 in.
Case of 6 for $430.47
Add to cart
 
Description

Description

Tychem™ CPF3 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. It is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Coverall with laydown collar and taped seam
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist and ankle
  • Commonly used in domestic preparedness for situations where potential to exposure to chemicals exist
Applications

Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities and domestic preparedness

Specifications

Specifications

Tychem™ CPF3 fabric
Tan
Taped
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit