Learn More
DuPont™ DuPont™ Tychem™ CPF 3 Coverall With Standard Fit Hood , Elastic Wrist and Ankles
Taped seams provide strong chemical resistance against heavy liquid splashes
Manufacturer:  DuPont™ C3127TTN4X000600
 View more versions of this product
Catalog No. 19-067-012
Add to cart
Description
Description
Tychem™ CPF3 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. It is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Coverall with standard fit hood and taped seam
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist and ankle
  • Commonly used in domestic preparedness for situations where potential to exposure to chemicals exist
Applications
Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities and domestic preparedness
Specifications
Specifications
Safety
6 ft.4 in. to 6 ft.7 in.
Taped
33.5 in.
Tan
Tychem™ CPF3 fabric
4X-Large