Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ DuPont™ Tychem™ CPF 3 Coverall With Standard Fit Hood , Elastic Wrist and Ankles

Taped seams provide strong chemical resistance against heavy liquid splashes

$438.88 - $511.52

Specifications

Material Tychem™ CPF3 fabric
Seam Type Taped
Color Tan
View More Specs

Products 3
Catalog Number Mfr. No. Size Width (English) Chest Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Width (English) Chest Height (English) Price Quantity & Availability  
19-067-011
View Documents
DuPont™
C3127TTN3X000600
3X-Large 31.25 in. 6 ft.2 in. to 6 ft. 4 in.
Case of 6 for $438.88
Add to cart
 
19-067-012
View Documents
DuPont™
C3127TTN4X000600
4X-Large 33.5 in. 6 ft. 4 in. to 6 ft. 7 in.
Case of 6 for $473.62
Add to cart
 
19-067-013
View Documents
DuPont™
C3127TTN5X000600
5X-Large 36 in. 6 ft. 7 in. to 6 ft. 10 in.
Case of 6 for $511.52
Add to cart
 
Description

Description

Tychem™ CPF3 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. It is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Coverall with standard fit hood and taped seam
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist and ankle
  • Commonly used in domestic preparedness for situations where potential to exposure to chemicals exist
Applications

Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities and domestic preparedness

Specifications

Specifications

Tychem™ CPF3 fabric
Taped
Tan
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit