Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem ™ 5000 Coveralls with Hood and Overboots/Socks

Protection against chemical splash as well as potential exposure to chemical warfare agents

Manufacturer:  DuPont™ C3122TTN2X0006BN

 View more versions of this product

Catalog No. 19-715-257


Add to cart

Alert:

Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.


Description

Description

Provide defense for applications such as acid handling, tank cleaning, chemical handling, petrochemical market, hazardous waste cleanup, industrial hazmat teams and fire department and utilities use.

  • Zipper front with attached hood and elastic wrists and overboots
  • Multilayer barrier film laminated to a lasting 2 oz. per sq. yd.
  • Polypropylene substrate offers greater physical strength and chemical holdout
  • Set sleeve pattern for greater arm movement
  • Coveralls have front-entry zipper
  • Tan

†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Compliance

Meets ANSI™/ISEA™ 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standard Methods for Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.

Specifications

Specifications

Attached socks
Safety
Zipper with storm flap
Elastic
Yes
6 to 6.4 ft.
Attached, Elastic face
93.98 cm
Taped
45.5 to 49 in.
115.57 to 124.46 cm
142.24 cm
0.27 lb.
0.12 kg
Tychem™ CPF 3
ASTM D1117, Berry Amendment
Tan
Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
1.82 to 1.95 m
37 in.
Hooded
2X-Large
30 in.
76.2 cm
0.45 mm
29.37 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit