DuPont ™ Tychem ™ CPF ™ 3 Coveralls with Hood and Overboots/Socks

Protection against chemical splash as well as potential exposure to chemical warfare agents

Manufacturer: Dupont Personal Protection C3122TTNMD000600

 View more versions of this product

Catalog No. 19-088-1230B

  Check Availability
Add to Cart

Alert

Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.


    Specifications

    Certifications/Compliance ASTM D1117, D5597, D5034 (Standard)
    Material Tychem™ CPF 3
    Color Tan
    Height (Metric) 1.6 to 1.73m
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Length (Metric) Sleeve 83.82cm
    Size (English) Inseam 28 in.
    Thickness 0.45mm
    Weight (English) 0.27 lb.
    Construction Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
    Includes Elastic around the face, tighter fit elastic wrist
    Seam Type Taped
    Size Medium
    Disposable Yes
    Size (Metric) Chest 85.09 to 93.98cm
    Height (English) 5.3 to 5.7 ft.
    Hood Type Attached
    Length (English) Sleeve 33 in.
    Closure Type Zipper, Storm Flap
    Size (Metric) Waist 116.84cm
    Width (English) Chest 23.37 in.
    Weight (Metric) 0.12kg
    Ankle Style Socks boots
    Cuff Style Elastic

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit