DuPont ™ Tychem ™ CPF ™ 3 Coveralls with Hood and Overboots/Socks

Protection against chemical splash as well as potential exposure to chemical warfare agents

Manufacturer: Dupont Personal Protection C3122TTNSM0006BN

 View more versions of this product

Catalog No. 19-715-253

  Check Availability
Add to Cart

Alert

Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.


    Specifications

    Certifications/Compliance ASTM D1117, Berry Amendment
    Color Tan
    Height (Metric) 1.52 to 1.73m
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Size (English) Inseam 27.5 in.
    Thickness 0.45mm
    Weight (English) 0.27 lb.
    Construction Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
    Ankle Style Attached socks
    Material Berry Amendement Compliant Tychem™ CPF 3
    Seam Type Taped
    Size Small
    Disposable Yes
    Hood Type Attached
    Length (English) Sleeve 32 in.
    Length (Metric) Sleeve 81.28cm
    Closure Type Zipper, Storm Flap
    Size (Metric) Inseam 69.85cm
    Size (Metric) Waist 113.03cm
    Width (English) Chest 21.37 in.
    Weight (Metric) 0.12kg
    Includes Elastic around the face, tighter fit elastic wrist
    Size (English) Chest 29.5 to 33 in.
    Size (Metric) Chest 74.93 to 83.82cm
    Height (English) 5 to 5.7 ft.
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit