Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
DuPont ™ Tychem ™ CPF ™ 3 Coveralls with Hood and Overboots/Socks
Protection against chemical splash as well as potential exposure to chemical warfare agents
Manufacturer: Dupont Personal Protection C3122TTN3X0006BN
 View more versions of this product
Catalog No. 19-715-258
  N/A
Add to Cart
Alert
Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.
Related Products
Product Content Feedback