DuPont ™ Tychem ™ CPF ™ 3 Coveralls with Respirator Fit Hoods, Berry Ammendment Compliant

Designed for use during rigorous activities

Manufacturer: Dupont Personal Protection C3128TTN2X0006BN

 View more versions of this product

Catalog No. 19-715-273

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D1117, Berry Amendment
    Material Tychem™ CPF 3
    Color Tan
    Height (Metric) 1.82 to 1.95m
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Size (English) Inseam 29.75 in.
    Thickness 0.45mm
    Weight (English) 0.27 lb.
    Construction Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
    Includes Elastic around the face, tighter fit elastic wrist
    Seam Type Taped
    Size 2X-Large
    Size (English) Chest 46.75 to 50.25 in.
    Disposable Yes
    Hood Type Attached
    Length (English) Sleeve 38 in.
    Length (Metric) Sleeve 96.52cm
    Closure Type Zipper, Storm Flap
    Size (Metric) Inseam 75.56cm
    Size (Metric) Waist 142.24cm
    Width (English) Chest 30 in.
    Weight (Metric) 0.12kg
    Ankle Style Attached socks/boots flaps
    Cuff Style Elastic
    Size (Metric) Chest 118.74 to 127.63cm
    Height (English) 6 to 6.4 ft.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit