DuPont ™ Tychem ™ CPF ™ 3 Coveralls with Respirator Fit Hoods, Berry Ammendment Compliant

Designed for use during rigorous activities

Manufacturer: Dupont Personal Protection C3128TTN3X0006BN

 View more versions of this product

Catalog No. 19-715-274

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D1117, Berry Amendment
    Material Tychem™ CPF 3
    Includes Elastic around the face, tighter fit elastic wrist
    Seam Type Taped
    Color Tan
    Size 3X-Large
    Size (English) Chest 50.75 to 54.25 in.
    Disposable Yes
    Size (Metric) Chest 128.9 to 137.79cm
    Height (English) 6.2 to 6.4 ft.
    Height (Metric) 1.88 to 1.95m
    Hood Type Attached
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Length (English) Sleeve 39 in.
    Length (Metric) Sleeve 99.06cm
    Closure Type Zipper, Storm Flap
    Size (English) Inseam 30.75 in.
    Size (Metric) Inseam 78cm
    Size (Metric) Waist 151.13cm
    Thickness 0.45mm
    Weight (English) 0.27 lb.
    Width (English) Chest 32 in.
    Weight (Metric) 0.12kg
    Construction Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
    Ankle Style Attached socks/boots flaps
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit