DuPont ™ Tychem ™ CPF ™ 3 HD Personal-Protection Garments

Integrated gloves eliminate need for taping and provide improved protection

Manufacturer: Dupont Personal Protection C3184TTN2X000600

 View more versions of this product

Catalog No. 19-130-1024E

  Check Availability
Add to Cart

    Specifications

    Material Tychem™ CPF 3
    Color Tan
    Size 2X-Large
    Height (Metric) 1.82 to 1.95m
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Length (Metric) Sleeve 99.69cm
    Size (English) Inseam 30 in.
    Size (Metric) Inseam 76.2cm
    Width (English) Chest 30 in.
    Construction Multilayer film barrier laminated to Polypropylene substrate
    Includes Collar, attached dual layer gloves, attached socks with outer boot flaps
    Seam Type Taped
    Size (English) Chest 46.75 to 50.25 in.
    Size (Metric) Chest 118.74 to 127.63cm
    Height (English) 6 to 6.4 ft.
    Length (English) Sleeve 39.25 in.
    Closure Type Front zipper and storm flap with hook and loop

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit