DuPont ™ Tychem ™ CPF ™ 3 HD Personal-Protection Garments

Integrated gloves eliminate need for taping and provide improved protection

Manufacturer: Dupont Personal Protection C3185TTNMD000600

 View all options for this product

Catalog No. 19-130-1023B

  Check Availability
Add to Cart


    Closure Type Front zipper and storm flap with hook and loop
    Construction Multilayer film barrier laminated to Polypropylene substrate
    Material Tychem™ CPF 3
    Includes Respirator fit hood, attached dual layer gloves, attached socks with outer boot flaps
    Seam Type Taped
    Color Tan
    Size Medium
    Size (English) Chest 34.75 to 38.25 in.
    Size (Metric) Chest 88.26 to 97.15cm
    Height (English) 5.3 to 5.7 ft.
    Height (Metric) 1.6 to 1.73m
    For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
    Length (English) Sleeve 35.25 in.
    Length (Metric) Sleeve 89.53cm
    Size (English) Inseam 28 in.
    Size (Metric) Inseam 71.12cm
    Width (English) Chest 24 in.

    Garments deliver proven barrier protection against 110 toxic industrial chemicals and are designed to stand up to chemical warfare agents including Sarin, Mustard, and Lewisite † Choose from two styles: with three-panel respirator-fitted hood or with collar for compatibility with hooded PAPR.

    • Tactile glove integrated into garment†—Stretch Chloroprene with Ansell™ Barrier™ inner glove for wide spectrum chemical protection
    • Heavy-duty front zipper closure and double storm flaps with wide hook and loop closure promote improved performance
    • Bigger zipper pull ensures easier donning and doffing
    • Includes attached socks‡ with boot splash cover
    • Taped seams
    • Light tan

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit