DuPon™ Tychem™ F Coveralls
Strong and lightweight, with broad chemical barrier
Manufacturer: Dupont Personal Protection TF169TGYXL0006US
 View more versions of this product
Catalog No. 19-130-5296
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance Standard
    Closure Type Zipper
    Construction Multilayer composite barrier film laminated to a Tyvek™ protective Fabric
    Material Tychem
    Includes Respirator fit hood, elastic wrist, attached socks, storm flap
    Seam Type Taped
    Color Gray
    Size X-Large
    For Use With (Application) Use when potential exposure to chemical warfare agents exists, widely used by military first Responder™ for WMD situations
    Use when potential exposure to chemical warfare agents exists; widely used by military first responders for WMD situations.
    • Taped seams
    • Attached respirator-fit hood with elastic around face
    • Front-zipper closure
    • Storm flap with double-sided tape over zipper
    • Elastic back
    • Elastic wrists
    • Gray
    • See www.fishersci.com for DuPont Tychem F Chemical Chart
  • Description & Specifications
Related Products
Product Content Feedback