Learn More
DuPont™ Tychem™ F Taped Seam Coveralls With Attached Socks (TAA Compliant)
Widely used by military personnel and first responders for chemical warfare situations
Manufacturer:  Dupont Personal Protection TF169TGY2X0006TV
 View more versions of this product
Catalog No. 19-162-815
  Check Availability
Add to cart
Specifications
Description
  • Strong and durable coverall with broad chemical barrier
  • Tychem™ F coverall is made of barrier film laminated to Tyvek™ substrate
  • For use when potential exposure to industrial chemicals and chemical warfare agents exists
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Available in low-visibility gray fabric for discretionary purposes
  • With taped seams and attached hood (respirator fit)
  • Front zipper closure and storm flap with tape closure
  • Elastic wrists and elastic ankles
Compliance
TAA Compliant
Description & Specifications
Specifications
Tychem™ F
Taped
Gray
2X-Large