DuPont ™ Tychem ™ LV Coveralls with Hood

Broadest range of chemical protection, with barrier data on over 285 chemicals

Manufacturer: Dupont Personal Protection LV128TODSM000200

 View all options for this product

Catalog No. 19-715-291


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Closure Type Zipper
    Construction Multilayer barrier films laminated to a high strength Polypropylene substrate
    Material Tychem
    Includes Respirator fit hood, elastic wrist, attached socks with outer boot flaps
    Seam Type Taped
    Color Olive Drab
    Size Small
    Size (English) Chest 32.25 to 35.75 in.
    Size (Metric) Chest 81.91 to 90.8cm
    Height (English) 5 to 5.7 ft.
    Height (Metric) 1.52 to 1.73m
    For Use With (Application) Military situations, chemical, biological terrorism incident response, chemical handling, hazardous materials, waste cleanup, fire departments and industrial hazmat teams
    Length (English) Sleeve 33 in.
    Length (Metric) Sleeve 83.82cm
    Size (English) Inseam 27.25 in.
    Size (Metric) Inseam 69.21cm
    Width (English) Chest 22.75 in.

    • Tychem LV — a multi-layer barrier film laminated to a high strength, 2.3 oz./yd2 polypropylene substrate
    • Fabric is strong, abrasion and puncture resistant, lightweight and flexible
    • Seams are both sewn and taped to provide strong chemical resistance against heavy liquid splashes and tough seam stress
    • Sewn seam is covered with a strip of compatible material by heat-sealing
    • Polyester front zipper closure with storm flap; flap has sticky tape closure
    • With attached hood
    • Color: Military Olive Drab
    Style LV127T (Without Boots)
    • Features elastic wrists and ankles
  • Style LV128T (With Attached Socks)
    • Includes elastic wrists
    • Respirator fit closure
    • Socks with outer boot flaps

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit