DuPont ™ Tychem ™ LV Coveralls with Hood

Broadest range of chemical protection, with barrier data on over 285 chemicals

Manufacturer: Dupont Personal Protection LV127TOD3X000200

 View all options for this product

Catalog No. 19-100-410


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Closure Type Zipper
    Construction Multilayer film barrier laminated to Polypropylene substrate
    Material Tychem
    Includes Standard fit hood, elastic wrist and ankle
    Seam Type Taped
    Color Military Olive Drab
    Size 3X-Large
    Size (English) Chest 47.25 to 50.75 in.
    Size (Metric) Chest 120 to 128.9cm
    Height (English) 6.2 to 6.4 ft.
    Height (Metric) 1.88 to 1.95m
    For Use With (Application) Military situations, chemical, biological terrorism incident response, chemical handling, hazardous materials, waste cleanup, fire departments and industrial hazmat teams
    Length (English) Sleeve 39.25 in.
    Length (Metric) Sleeve 99.69cm
    Size (English) Inseam 31 in.
    Width (English) Chest 30.25 in.

    • Tychem LV — a multi-layer barrier film laminated to a high strength, 2.3 oz./yd2 polypropylene substrate
    • Fabric is strong, abrasion and puncture resistant, lightweight and flexible
    • Seams are both sewn and taped to provide strong chemical resistance against heavy liquid splashes and tough seam stress
    • Sewn seam is covered with a strip of compatible material by heat-sealing
    • Polyester front zipper closure with storm flap; flap has sticky tape closure
    • With attached hood
    • Color: Military Olive Drab
    Style LV127T (Without Boots)
    • Features elastic wrists and ankles
  • Style LV128T (With Attached Socks)
    • Includes elastic wrists
    • Respirator fit closure
    • Socks with outer boot flaps

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit