Learn More
DuPont ™ Tychem ™ LV Coveralls with Hood
Broadest range of chemical protection, with barrier data on over 285 chemicals
Manufacturer:  Dupont Personal Protection LV127TODXL000200
 View more versions of this product
Catalog No. 19-100-413
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Tychem™ fabrics offer little to no thermal heat protection to the user.
Specifications Description & Specifications
Specifications
Zipper
Tychem
Taped
X-Large
104.77 to 113.66cm
1.76 to 1.88m
37.25 in.
29 in.
Multilayer film barrier laminated to Polypropylene substrate
Standard fit hood, elastic wrist and ankle
Military Olive Drab
41.25 to 44.75 in.
5.8 to 6.2 ft.
Military situations, chemical, biological terrorism incident response, chemical handling, hazardous materials, waste cleanup, fire departments and industrial hazmat teams
94.61cm
27.25 in.