DuPont ™ Tychem ™ LV One-Piece Suits

Manufacturer: Dupont Personal Protection LV611TODXL000100

 View more versions of this product

Catalog No. 19-066-108

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance NFPA 1992, NFPA 1994, Class 2
    Color Olive Green
    Height (Metric) 1.76 to 1.88m
    For Use With (Application) Military situations, chemical, biological terrorism incident response, chemical handling, hazardous materials, waste cleanup, fire departments and industrial hazmat teams
    Material Tychem™ LV
    Includes SCBA interface hood, rear entry, attached dual layer gloves, multilayer laminate / outer, Neoprene, glove size 10, attached socks with outer boot flaps, triple storm flap with hook and loop closure
    Seam Type Taped
    Size X-Large
    Size (English) Chest 42.75 to 46.25 in.
    Size (Metric) Chest 108.58 to 117.47cm
    Height (English) 5.8 to 6.2 ft.
    Length (English) Sleeve 38 in.
    Length (Metric) Sleeve 96.52cm
    Closure Type Hook and loop
    Size (English) Inseam 29.75 in.
    Size (Metric) Inseam 75.56cm
    Thickness 18mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit