Learn More
DuPont ™ Tychem ™ Reflector ™ Chemical Suit, NFPA™ 1991 2000 Edition
Limited-use garment that combines broad chemical holdout with flash-fire protection
Manufacturer:  Dupont Personal Protection RF600TSVLG000100
 View more versions of this product
Catalog No. 18-999-336B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1991
Tychem™ Reflector™
Triple taped
Large
121.28 to 130.75cm
1.79 to 1.92m
29 in.
31.5 in.
40mil
Zipper front
Standard width faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal North Silver Shield™/4H™/external Neoprene size 10 gloves, separate Kevlar™ knit over gloves, socks with outer boot flaps, soles
Silver
47.75 to 51.25 in.
5.9 to 6.3 ft.
Ideal for industrial chemical, HazMat response and military applications
73.66cm
80cm
Accessories