DuPont™ Tychem™ SL 122 Series Coveralls, Berry Amendment Compliant

Delivers effective protection against a range of chemical environments.

Manufacturer: DuPont™ SL122TWHXL0006BN

 View more versions of this product

Catalog No. 19-162-776

  Check Availability
Add to Cart

Alert

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Material Saranex 23-P film laminated to Tyvek fabric
    Includes Attached hood, elastic wrist, attached socks
    Disposable Yes
    Height (English) 5.66 to 6.16 ft.
    Hood Type Attached hood with elastic around face opening
    For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
    Closure Type Front zipper
    Sterile No
    Width (English) Chest 27.25 in.
    Width (Metric) Chest 69.21cm
    Size (English) Chest 41.25 to 44 .75 in.
    Height (Metric) 1.70 to 1.85m
    Length (English) Sleeve 36.5 in.
    Length (Metric) Sleeve 92.71cm
    Size (English) Inseam 29 in.
    Size (Metric) Inseam 73.66cm
    Thickness 12mil
    Cuff Style Elastic
    Product Type Coverall with attached hood and socks
    Seam Type Taped
    Color White
    Size X-Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit