Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ SL 128 Series Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
4X-Large
5X-Large
Description
DuPont Tychem 4000 delivers effective protection against a range of chemical environments. Tychem 4000, utilizes Saranex 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion. Tychem 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face maskStrong zipper for tough, flexible closureStorm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipperElastic opening for tighter fit at wristIntegrated socks composed of garment materialAttached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Size | Large |
Material | Tychem |
Color | White |
Ankle Style | Attached Socks |
Neck Style | Hooded |
Hood Type | Attached |
Closure Type | Front Zipper |
Seam Type | Taped |
For Use With (Application) | Waste management, hazardous response and nuclear,environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction