DuPont™ Tychem™ SL 128 Series Coveralls

Manufacturer: Dupont Personal Protection SL128TWHMD000600

 View more versions of this product

Catalog No. 19-162-785

  Check Availability
Add to Cart

    Specifications

    Material Tychem™ SL
    Includes Attached hood and socks
    Seam Type Taped
    Color White
    Size Medium
    Hood Type Attached
    For Use With (Application) Waste management, hazardous response and nuclear,environments
    Closure Type Front zipper closure

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit