Learn More
DuPont™ Tychem™ SL Sleeved Apron
Made of Tyvek™ spunbonded olefin laminated with Saranex™-23P to provide chemical barrier
Manufacturer:  Dupont Personal Protection SL275TWH2X002500
 View more versions of this product
Catalog No. 19-715-340
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
44 in.
28 in.
White
Protective Apron with Snap and Ties
Tyvek™
10.0
12mil
111.7cm
71.12cm
Elastic
2X-Large
44 x 28 in. (111.7 x 71.12cm)
Taped