Learn More
DuPont™ Tychem™ SL Apron
Made of Tyvek™ spunbonded olefin laminated with Saranex™-23P to provide chemical barrier
$448.00 - $504.00
Specifications
Closure Type Snap and Ties
Cuff Style Elastic
Color White
Material Tyvek™
View More Specs
Products 5
Catalog Number Mfr. No. Size Seam Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Seam Type Price Quantity & Availability  
19-715-338
View Documents
DuPont™
SL275TWHLG002500
Large Taped
Case of 25 for $448.00
Add to cart
19-715-341
View Documents
DuPont™
SL275TWH3X002500
3X-Large Taped
Case of 25 for $504.00
Add to cart
19-715-340
View Documents
DuPont™
SL275TWH2X002500
2X-Large Taped
Case of 25 for $475.00
Add to cart
19-715-337
View Documents
DuPont™
SL275TWHMD002500
Medium Bound
Case of 25 for $448.00
Add to cart
19-715-339
View Documents
DuPont™
SL275TWHXL002500
X-Large Taped
Case of 25 for $448.00
Add to cart
Description
Description
  • Knee-length apron/gown design with attached sleeves
  • Provides enhanced frontal protection
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • Sewn seam is covered with strip of compatible chemical-resistant material through heat sealing
  • Open back for wearer comfort
  • Bound seams around neck opening with snap closure
  • Ties at waist
  • Elastic opening for tighter fit at wrist
  • 44 in. long for neck-to-knee coverage
  • Available in multiple sizes
  • Color: White
Order Info
See chemical permeation data chart on this fabric.
Specifications
Specifications
Snap and Ties
Elastic
White
Tyvek™