Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ SL Coveralls with Elastic Respirator Fit Closure
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Height (English):
6 to 6.4 ft.
6.2 to 6.4 ft.
Height (Metric):
1.82 to 1.95 m
1.88 to 1.95 m
Description
Used in environmental clean-up operations, waste management, industrial plants, cleanroom applications, hazardous material response teams, and emergency services.
- DuPont™ Tyvek ™ fabric laminated with Dow™ Saranex™-23P film
- All models fitted with attached hoods, elastic respirator fit closures, elastic wrists, and front zipper closures with storm flaps
- 19-822 Series has attached sock boots†, and boot flaps
- 19-130-4 Series has attached boots
- 19-130-1 Series has attached boots and is manufactured in the U.S.
Order Info
19-130-1 Seriesis manufactured in the U.S.
†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.
Specifications
Size | X-Large |
Material | Tychem™ |
Color | White |