DuPont ™ Tychem ™ SL Coveralls with Hoods

Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments

Manufacturer: Dupont Personal Protection SL127BWHLG001200

 View more versions of this product

Catalog No. 18-643

  Check Availability
Add to Cart

Alert

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Certifications/Compliance ASTM
    Color White
    For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
    Thickness 12mil
    Weight (English) 0.22 lb.
    Material Tychem™ SL
    Includes Coveralls with hood, elastic wrist, ankle
    Seam Type Bound
    Size Large
    Size (Metric) Chest 97.15 to 106.04cm
    Height (English) 5.5 to 5.9 ft.
    Height (Metric) 1.67 to 1.8m
    Length (English) Sleeve 35 in.
    Length (Metric) Sleeve 88.9cm
    Closure Type Zipper
    Size (English) Inseam 28.5 in.
    Width (English) Chest 25.75 in.
    Weight (Metric) 0.1kg
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit