DuPont ™ Tychem ™ ThermoPro Chemical Protection Aprons

A single layer garment that provides triple hazard protection from flash fire, electric arc and liquid-chemical splash

Manufacturer: Dupont Personal Protection TP278TGY00000400

 View more versions of this product

Catalog No. 19-120-955

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Material Tychem™ ThermoPro
    Thickness (English) 34mil
    Thickness (Metric) 0.86mm
    Width (Metric) 71.7cm
    Width (English) 28.25 in.
    Color Gray
    Seam Type Bound
    Certifications/Compliance NFPA 1992 (2005) and NFPA 2112 (2007)
    Dimensions (L x W) 52 x 28.25 in. (132.08 x 71.7cm)
    Length (English) 52 in.
    Length (Metric) 132.08cm
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit