Learn More
DuPont™ Tychem™ ThermoPro Combo Suits
Protection from chemicals, flash fire and electrical arc
$814.38 - $988.42
Specifications
Ankle Style Open
Collar Type Mandarin
Cuff Style Elastic
View More Specs
Includes:
  • Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
  • Bib Overall with taped seams and adjustable webbing straps with closure
Products 14
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Color Price Quantity & Availability  
19-120-964
View Documents Promotion Details
DuPont™
TP750TORSM000200
Small Orange
Case of 2 for $814.38
Add to cart
19-120-965
View Documents Promotion Details
DuPont™
TP750TORMD000200
Medium Orange
Case of 2 for $814.38
Add to cart
19-120-966
View Documents Promotion Details
DuPont™
TP750TORLG000200
Large Orange
Case of 2 for $814.38
Add to cart
19-120-967
View Documents Promotion Details
DuPont™
TP750TORXL000200
X-Large Orange
Case of 2 for $814.38
Add to cart
19-120-970
View Documents Promotion Details
DuPont™
TP750TOR4X000200
4X-Large Orange
Case of 2 for $988.42
Add to cart
19-120-969
View Documents Promotion Details
DuPont™
TP750TOR3X000200
3X-Large Orange
Case of 2 for $915.08
Add to cart
19-120-968
View Documents Promotion Details
DuPont™
TP750TOR2X000200
2X-Large Orange
Case of 2 for $863.64
Add to cart
19-120-957
View Documents Promotion Details
DuPont™
TP750TGYSM000200
Small Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-959
View Documents Promotion Details
DuPont™
TP750TGYLG000200
Large Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-960
View Documents Promotion Details
DuPont™
TP750TGYXL000200
X-Large Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-961
View Documents Promotion Details
DuPont™
TP750TGY2X000200
2X-Large Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-958
View Documents Promotion Details
DuPont™
TP750TGYMD000200
Medium Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-963
View Documents Promotion Details
DuPont™
TP750TGY4X000200
4X-Large Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-120-962
View Documents Promotion Details
DuPont™
TP750TGY3X000200
3X-Large Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Constructed for heavy use, yet lightweight and easy to wear
  • At least 30 minutes of protection against more than 180 chemical challenges
  • Arc rating of 15.22 cal/cm Ebt
Specifications
Specifications
Open
Mandarin
Elastic