Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
Click to view available options
Certifications/Compliance:
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Color:
Gray
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6.7 to 6.10 ft.
Description
ThermoPro coveralls are secure in fit to seal out contaminants
- Respirator fit hood with drawstrings
- Front zipper with storm flap, hook and loop secure
- Elastic wrists
- Open, hemmed ankles
- Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
- At least 30 minutes of protection against 182 chemical challenges
- Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
- HazMat teams in fire departments
- Industrial fire brigades
- Clandestine lab investigation
- Industrial chemical processing plants
Certifications
- NFPA 1992 (2012)
- NFPA 2112 (2012)
- Exceeds NFPA 70E Hazard Risk Category 2
Specifications
Size | Medium |
Material | HDPE |
Color | Gray |
Ankle Style | Open |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Hook and Loop |
Seam Type | Taped |
Certifications/Compliance | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Description | Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction