Learn More
DuPont™ Tychem™ TK 128T Coveralls
Offers protection from a vast array of chemicals including toxic and corrosive agents
Manufacturer:  DuPont™ TK1227/DEL
 View more versions of this product
Catalog No. 19-120-2076G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
Description
These coveralls features strong, durable material and several measures to seal out chemicals.
  • Tychem TK fabric
  • Extremely durable, puncture- and tear-resistant
  • Respirator fit hood
  • Strong zipper, flexible closure, storm flaps with adhesive seal
  • Attached socks
  • Attached boot covers
  • Elastic wrists
  • Protection against corrosive, toxic gases, liquid and solid chemicals
  • Industrial, HazMat, and domestic preparedness
Compliance
Trade Agreement Act Compliant
Specifications
Specifications
Lime
4X-Large
Tychem