DuPont ™ Tychem ™ TK Level A Suits

Manufacturer: Dupont Personal Protection TK555TLYLG00015C

 View more versions of this product

Catalog No. 19-121-832C

  Check Availability
Add to Cart

    Specifications

    Material Tychem™ TK
    Includes Expanded Back; Knee wear pads; Exta wide 3-layer faceshield: PVC 40mil/PTFE 5mil/PVC 20mil; w/viton gloves
    Seam Type Double taped
    Color Lime yellow
    Size Large
    For Use With (Application) Industrial, HazMat and domestic preparedness applications
    Closure Type Gas-tight PVC zipper closure and double storm flaps with hook-and-loop closure
    Style Rear entry
    Construction Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit