Learn More
DuPont™ Tyvek™ 127S Series Coveralls
Manufacturer:  DuPont™ TY127SWH2X002500
 View more versions of this product
Catalog No. 01-361-7E
Add to cart
Description
Description
Tyvek 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. The fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
Specifications
Specifications
45.25 to 48.75 in.
Elastic
Hooded
Zipper front
Lay down
Serged
2X-Large
1.82 to 1.95m
38.5 in.
31 in.
5.7mil
114.93 to 123.82cm
Yes
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
HDPE
Elastic
White
6 to 6.4 ft.
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
97.79cm
78.74cm
Documents
Accessories