Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ Series 122S Coveralls

Comfort fit design based on extensive wearer input to provide a comfortable garment design that enables a greater range of movement while stretching and bending.

Manufacturer:  DuPont™ TY122SWH5X002500

 View more versions of this product

Catalog No. 01-361-2H


Add to cart

Alert:

Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.


Description

Description

Comfort fit design based on extensive wearer input to provide a comfortable garment design that enables a greater range of movement while stretching and bending.

  • Comfort fit design provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Attached Tyvek FC boots have special coating to provide added skid-resistance
  • Longer zipper extends to chin for complete coverage of neck area and provides easeir donning and doffing
  • Elastic opening for tighter fit at wrist
  • Elastic waist to better position garment
  • Elastic ankles
  • Serged seam
  • White
Specifications

Specifications

Elastic
White
6.7 to 6.10 ft.
Respirator-fit
39.5 in.
33.5 in.
5.9 mils
Elastic
Zipper front
Hooded
Attached skid-resistant boots
141.6 to 150.5cm
Serged
5X-Large
2.0 to 1.8m
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
100.3cm
85cm
34.5 in.
87.6
Elastic, Attached boots
Yes
Flash spun high-density polyethylene
55.75 to 59.25 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit