Learn More
DuPont Tyvek 121S Series Coveralls
Provide excellent inherent protection against submicron airborne particles
$271.00 - $355.00
Specifications
Collar Type | Lay down |
---|---|
Closure Type | Front zipper |
Color | White |
Cuff Style | Elastic |
Description | Attached skid-resistant boots |
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Size | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Material | Price | Quantity & Availability | ||||
19-100-377
|
DuPont
TY121SWHMD0025NS |
Medium | Flashspun HDPE |
Case of 25 for $271.00
|
|
19-100-380
|
DuPont
TY121SWHLG0025NS |
Large | Spun HDPE |
Case of 25 for $271.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-100-378
|
DuPont
TY121SWHXL0025NS |
X-Large | Spun HDPE |
Case of 25 for $271.00
|
|
19-100-379
|
DuPont
TY121SWH2X0025NS |
2X-Large | Spun HDPE |
Case of 25 for $287.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-100-382
|
DuPont
TY121SWH3X0025NS |
3X-Large | Spun HDPE |
Case of 25 for $304.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-100-482
|
DuPont
TY121SWH4X0025NS |
4X-Large | Flashspun HDPE |
Case of 25 for $329.00
|
|
19-100-483
|
DuPont
TY121SWH5X0025NS |
5X-Large | Spun HDPE |
Case of 25 for $355.00
|
|
Description
Tyvek fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
- Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Laydown collar
- Elastic opening for tighter fit at wrist
- Elastic ankles
- Serged seams
- Front zipper closure
- White
- Attached Tyvek FC boots have special coating to provide added skid-resistance
Compliance
Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
Specifications
Lay down | |
White | |
Attached skid-resistant boots | |
Yes |
Front zipper | |
Elastic | |
Serged | |
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.